Drukuj

Anti-HDAC3 Mouse Monoclonal Antibody [clone: 13B370]

Dostawca: United States Biological
USBI030292EA 2850 PLN
USBI030292
Anti-HDAC3 Mouse Monoclonal Antibody [clone: 13B370]
Przeciwciała
Type: Primary
Antigen: HDAC3 (histone deacetylase 3)
Clonality: Monoclonal
Clone: 13B370
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR