Drukuj
Anti-HFE Mouse Monoclonal Antibody [clone: 13B375]
Dostawca: United States Biological
USBI030297EA 2960 PLN
USBI030297
Anti-HFE Mouse Monoclonal Antibody [clone: 13B375]
Przeciwciała
Type: Primary
Antigen: HFE (hemochromatosis)
Clonality: Monoclonal
Clone: 13B375
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human