Drukuj

Anti-HIF1A Mouse Monoclonal Antibody [clone: 13B376]

Dostawca: United States Biological
USBI030298EA 2960 PLN
USBI030298
Anti-HIF1A Mouse Monoclonal Antibody [clone: 13B376]
Przeciwciała
Type: Primary
Antigen: HIF1A (hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor))
Clonality: Monoclonal
Clone: 13B376
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR