Drukuj

Anti-HPRT1 Mouse Monoclonal Antibody [clone: 13B381]

Dostawca: United States Biological
USBI030303EA 2540 PLN
USBI030303
Anti-HPRT1 Mouse Monoclonal Antibody [clone: 13B381]
Przeciwciała
Type: Primary
Antigen: HPRT1 (hypoxanthine phosphoribosyltransferase 1)
Clonality: Monoclonal
Clone: 13B381
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR