Drukuj

Anti-HSPB1 Mouse Monoclonal Antibody [clone: 13B384]

Dostawca: United States Biological
USBI030306EA 2960 PLN
USBI030306
Anti-HSPB1 Mouse Monoclonal Antibody [clone: 13B384]
Przeciwciała
Type: Primary
Antigen: HSPB1 (heat shock 27kDa protein 1)
Clonality: Monoclonal
Clone: 13B384
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR