Drukuj

Anti-HSPD1 Mouse Monoclonal Antibody [clone: 13B385]

Dostawca: United States Biological
USBI030307EA 2960 PLN
USBI030307
Anti-HSPD1 Mouse Monoclonal Antibody [clone: 13B385]
Przeciwciała
Type: Primary
Antigen: HSPD1 (heat shock 60kDa protein 1 (chaperonin))
Clonality: Monoclonal
Clone: 13B385
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR