Drukuj
Anti-HSP90AB1 Mouse Monoclonal Antibody [clone: 13B388]
Dostawca: United States Biological
USBI030310EA 2960 PLN
USBI030310
Anti-HSP90AB1 Mouse Monoclonal Antibody [clone: 13B388]
Przeciwciała
Type: Primary
Antigen: HSP90AB1 (heat shock protein 90kDa alpha (cytosolic), class B member 1)
Clonality: Monoclonal
Clone: 13B388
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Monkey, Mouse, Rat