Drukuj

Anti-LKAAEAR1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

USBI032896-FITCEA 2140 PLN
USBI032896-FITC
Anti-LKAAEAR1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50; Immunofluorescence: 1:10-50; Flow Cytometry: 1:10-50
Do not freeze!

Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50; Immunofluorescence: 1:10-50; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: LKAAEAR1 (LKAAEAR motif containing 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR