Drukuj
Anti-C2orf49 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Dostawca: US Biological
USBI032932-PEEA 4280 PLN
USBI032932-PE
Anti-C2orf49 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Przeciwciała
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50
Do not freeze!
Type: Primary
Antigen: C2orf49 (chromosome 2 open reading frame 49)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human