Drukuj

Anti-TEX37 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Dostawca: US Biological
USBI032934-APEA 4280 PLN
USBI032934-AP
Anti-TEX37 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: TEX37 (testis expressed 37)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR