Drukuj
Anti-PTCHD4 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Dostawca: US Biological
USBI033015-PEEA 4280 PLN
USBI033015-PE
Anti-PTCHD4 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Przeciwciała
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!
Type: Primary
Antigen: PTCHD4 (patched domain containing 4)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human