Drukuj

Anti-ERMARD Rabbit Polyclonal Antibody

Dostawca: US Biological
USBI033035EA 2950 PLN
USBI033035
Anti-ERMARD Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100; Flow Cytometry: 1:10-50

Type: Primary
Antigen: ERMARD (ER membrane-associated RNA degradation)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR