Drukuj
Anti-CCZ1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Dostawca: US Biological
USBI033043-APEA 4280 PLN
USBI033043-AP
Anti-CCZ1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!
Type: Primary
Antigen: CCZ1 (CCZ1 vacuolar protein trafficking and biogenesis associated homolog (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse