Drukuj

Anti-CCZ1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dostawca: US Biological
USBI033043-HRPEA 4280 PLN
USBI033043-HRP
Anti-CCZ1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: CCZ1 (CCZ1 vacuolar protein trafficking and biogenesis associated homolog (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR