Drukuj

Anti-YAE1D1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Dostawca: US Biological
USBI033046-PEEA 4280 PLN
USBI033046-PE
Anti-YAE1D1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Przeciwciała
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: YAE1D1 (Yae1 domain containing 1)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR