Drukuj

Anti-PPP1R35 Rabbit Polyclonal Antibody

Dostawca: US Biological
USBI033049EA 2950 PLN
USBI033049
Anti-PPP1R35 Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100

Type: Primary
Antigen: PPP1R35 (protein phosphatase 1, regulatory subunit 35)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR