Drukuj

Anti-C7orf63 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dostawca: US Biological
USBI033056-FITCEA 4280 PLN
USBI033056-FITC
Anti-C7orf63 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Przeciwciała
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!

Type: Primary
Antigen: C7orf63 (chromosome 7 open reading frame 63)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR