Drukuj
Anti-C7orf63 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Dostawca: US Biological
USBI033056-HRPEA 4280 PLN
USBI033056-HRP
Anti-C7orf63 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Type: Primary
Antigen: C7orf63 (chromosome 7 open reading frame 63)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human