Drukuj

Anti-C8B Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Dostawca: US Biological
USBI033058-APEA 4280 PLN
USBI033058-AP
Anti-C8B Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: C8B (complement component 8, beta polypeptide)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR