Drukuj

Anti-MKLN1 Rabbit Polyclonal Antibody

Dostawca: US Biological
USBI038453EA 2950 PLN
USBI038453
Anti-MKLN1 Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: MKLN1 (muskelin 1, intracellular mediator containing kelch motifs)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR