Drukuj
Anti-MKLN1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Dostawca: US Biological
USBI038453-APCEA 4280 PLN
USBI038453-APC
Anti-MKLN1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Przeciwciała
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: MKLN1 (muskelin 1, intracellular mediator containing kelch motifs)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human