Drukuj

Anti-MKNK2 Rabbit Polyclonal Antibody

Dostawca: US Biological
USBI038455EA 2950 PLN
USBI038455
Anti-MKNK2 Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: MKNK2 (MAP kinase interacting serine/threonine kinase 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR