Drukuj
Anti-MKRN2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Dostawca: US Biological
USBI038458-APCEA 4280 PLN
USBI038458-APC
Anti-MKRN2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Przeciwciała
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: MKRN2 (makorin ring finger protein 2)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human