Chcesz zalogować się jeszcze raz?
Anti-MKRN2 Rabbit Polyclonal Antibody (Biotin)
Dostawca: US Biological
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: MKRN2 (makorin ring finger protein 2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
ZAMÓW
Rozwiń / Zwiń
Rozmiar Ilość Numer katalogowy VWR Ilość
200 µl 200 µl USBI038458-BIOTIN
Wygeneruj kody kreskowe Format kodu kreskowego: 
Wygeneruj stronę katalogową PDF
SPECYFIKACJE
Antigen symbol MKRN2
Antigen name makorin ring finger Protein 2
Reactivity Human
Conjugation Biotin
Clonality Polyclonal
Antibody type Primary
Host Rabbit
Isotype IgG
Western blot Yes
ELISA Yes
Amino acid number 77 to 106
Epitope N-terminal
Purification purified by Protein A chromatography
Form liquid
Storage buffer PBS, pH 7,2
Temperatura przechowywania Cool
Shipping temperature Ice
Wprowadź szczegóły
Select the article for which you are looking for Certificate
Podaj numer partii. To pole jest obowiązkowe. Wpisz "X" dla VWR Chemicals, jeśli nr partii nie jest znany.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
albo
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Opis Numer katalogowy Availability Jednostka Twoja Cena Price Per Ilość
A new look and a renewed commitment
Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.
Learn More Start Shopping