Drukuj
Anti-MLC1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Dostawca: US Biological
USBI038461-PEEA 4280 PLN
USBI038461-PE
Anti-MLC1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Przeciwciała
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: MLC1 (megalencephalic leukoencephalopathy with subcortical cysts 1)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human