Drukuj
Anti-CENPU Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Dostawca: US Biological
USBI038466-FITCEA 4280 PLN
USBI038466-FITC
Anti-CENPU Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Przeciwciała
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: CENPU (centromere protein U)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human