Drukuj

Anti-MLH1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Dostawca: US Biological
USBI038468-APEA 4280 PLN
USBI038468-AP
Anti-MLH1 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50
Do not freeze!

Type: Primary
Antigen: MLH1 (mutL homolog 1)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR