Drukuj
Anti-MLXIP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Dostawca: US Biological
USBI038488-HRPEA 4280 PLN
USBI038488-HRP
Anti-MLXIP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Type: Primary
Antigen: MLXIP (MLX interacting protein)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse