Drukuj
Anti-MMAA Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Dostawca: US Biological
USBI038490-PEEA 4280 PLN
USBI038490-PE
Anti-MMAA Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Przeciwciała
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!
Type: Primary
Antigen: MMAA (methylmalonic aciduria (cobalamin deficiency) cblA type)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse