Drukuj

Anti-MMACHC Rabbit Polyclonal Antibody

Dostawca: US Biological
USBI038491EA 2950 PLN
USBI038491
Anti-MMACHC Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: MMACHC (methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR