Drukuj
Anti-MMACHC Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Dostawca: US Biological
USBI038491-APCEA 4280 PLN
USBI038491-APC
Anti-MMACHC Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Przeciwciała
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: MMACHC (methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human