Drukuj
Anti-MMACHC Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Dostawca: US Biological
USBI038491-HRPEA 4280 PLN
USBI038491-HRP
Anti-MMACHC Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: MMACHC (methylmalonic aciduria (cobalamin deficiency) cblC type, with homocystinuria)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human