Drukuj
Anti-MMD2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Dostawca: US Biological
USBI038494-HRPEA 4280 PLN
USBI038494-HRP
Anti-MMD2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Type: Primary
Antigen: MMD2 (monocyte to macrophage differentiation-associated 2)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse