Drukuj
Anti-MME Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Dostawca: US Biological
USBI038495-APEA 4280 PLN
USBI038495-AP
Anti-MME Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!
Type: Primary
Antigen: MME (membrane metallo-endopeptidase)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse