Drukuj

Anti-MMEL1 Rabbit Polyclonal Antibody

Dostawca: US Biological
USBI038496EA 2950 PLN
USBI038496
Anti-MMEL1 Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: MMEL1 (membrane metallo-endopeptidase-like 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR