Drukuj

Anti-OR5I1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Dostawca: US Biological
USBI039457-PEEA 4280 PLN
USBI039457-PE
Anti-OR5I1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Przeciwciała
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: OR5I1 (olfactory receptor, family 5, subfamily I, member 1)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR