Drukuj

Anti-OR5M1 Rabbit Polyclonal Antibody

Dostawca: US Biological
USBI039460EA 2950 PLN
USBI039460
Anti-OR5M1 Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: OR5M1 (olfactory receptor, family 5, subfamily M, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR