Drukuj
Anti-OR5M8 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Dostawca: US Biological
USBI039461-APEA 4280 PLN
USBI039461-AP
Anti-OR5M8 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: OR5M8 (olfactory receptor, family 5, subfamily M, member 8)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human