Drukuj

Anti-OR5M8 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dostawca: US Biological
USBI039461-FITCEA 4280 PLN
USBI039461-FITC
Anti-OR5M8 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Przeciwciała
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: OR5M8 (olfactory receptor, family 5, subfamily M, member 8)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR