Drukuj

Anti-OR6B2 Rabbit Polyclonal Antibody

Dostawca: US Biological
USBI039465EA 2950 PLN
USBI039465
Anti-OR6B2 Rabbit Polyclonal Antibody
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: OR6B2 (olfactory receptor, family 6, subfamily B, member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR