Drukuj
Anti-OR6B2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Dostawca: US Biological
USBI039465-FITCEA 4280 PLN
USBI039465-FITC
Anti-OR6B2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Przeciwciała
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: OR6B2 (olfactory receptor, family 6, subfamily B, member 2)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human