Drukuj

Anti-OR6C4 Rabbit Polyclonal Antibody (Biotin)

Dostawca: US Biological
USBI039468-BIOTINEA 4280 PLN
USBI039468-BIOTIN
Anti-OR6C4 Rabbit Polyclonal Antibody (Biotin)
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Flow Cytometry: 1:10-50

Type: Primary
Antigen: OR6C4 (olfactory receptor, family 6, subfamily C, member 4)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR