Drukuj

Anti-OR6K2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Dostawca: US Biological
USBI039470-APCEA 4280 PLN
USBI039470-APC
Anti-OR6K2 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Przeciwciała
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: OR6K2 (olfactory receptor, family 6, subfamily K, member 2)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR