Drukuj

Anti-OR6K2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Dostawca: US Biological
USBI039470-HRPEA 4280 PLN
USBI039470-HRP
Anti-OR6K2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: OR6K2 (olfactory receptor, family 6, subfamily K, member 2)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR