Drukuj
Anti-OR6K3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Dostawca: US Biological
USBI039471-APCEA 4280 PLN
USBI039471-APC
Anti-OR6K3 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Przeciwciała
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!
Type: Primary
Antigen: OR6K3 (olfactory receptor, family 6, subfamily K, member 3)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human