Drukuj

Anti-STX6 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Dostawca: US Biological
USBI042442-FITCEA 4280 PLN
USBI042442-FITC
Anti-STX6 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Przeciwciała
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: STX6 (syntaxin 6)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR