Drukuj

Anti-WARS Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Dostawca: United States Biological
USBI043801-APEA 4280 PLN
USBI043801-AP
Anti-WARS Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Przeciwciała
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: WARS (tryptophanyl-tRNA synthetase)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR