Drukuj

Anti-GITRL Rat Monoclonal Antibody [clone: MGTL 10]

Dostawca: United States Biological
USBI044849EA 1880 PLN
USBI044849
Anti-GITRL Rat Monoclonal Antibody [clone: MGTL 10]
Przeciwciała
Type: Primary
Antigen: GITRL
Clonality: Monoclonal
Clone: MGTL 10
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR