Drukuj

Anti-GITRL Rabbit Polyclonal Antibody

Dostawca: United States Biological
USBI044852EA 2210 PLN
USBI044852
Anti-GITRL Rabbit Polyclonal Antibody
Przeciwciała
Type: Primary
Antigen: GITRL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Zobacz ceny

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Więcej About VWR