Drukuj
Anti-SPA17 Mouse Monoclonal Antibody [clone: 3B6]
Dostawca: United States Biological
USBI133739EA 2620 PLN
USBI133739
Anti-SPA17 Mouse Monoclonal Antibody [clone: 3B6]
Przeciwciała
Type: Primary
Antigen: SPA17 (sperm autoantigenic protein 17)
Clonality: monoclonal
Clone: 3B6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human